4'-(Trifluormethyl)acetanilid

Lieferant: FLUOROCHEM

002700-5G 002700-25G
FLUO002700-5GEA 45.9 EUR
FLUO002700-5G FLUO002700-25G
4'-(Trifluormethyl)acetanilid
4'-(Trifluormethyl)acetanilid
Formel: C₉H₈F₃NO
Schmelzpunkt: 208…211 °C
MDL: MFCD00013562
CAS-Nummer: 349-97-3
EINECS: 206-495-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR