2,3,5-Trimethylpyrazin

Lieferant: FLUOROCHEM

002732-25G
FLUO002732-25GEA 37.1 EUR
FLUO002732-25G
2,3,5-Trimethylpyrazin
2,3,5-Trimethylpyrazine
Formel: C₇H₁₀N₂
Molecular Weight: 122.17 g/mol
Siedepunkt: 171
Dichte: 0.9958 g/cm³ (0 °C)
MDL: MFCD00006145
CAS-Nummer: 14667-55-1
EINECS: 238-712-0
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR