4-Methyloctansäure

Lieferant: FLUOROCHEM

002774-1G 002774-5G
FLUO002774-1GEA 30.6 EUR
FLUO002774-1G FLUO002774-5G
4-Methyloctansäure
4-Methylcaprylsäure
Formel: C₉H₁₈O₂
MDL: MFCD00051938
CAS-Nummer: 54947-74-9
EINECS: 259-404-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR