Cobalt(III)fluorid
Lieferant: FLUOROCHEM
|
Gefahr
|
002840-100G
002840-500G
FLUO002840-100GEA
422
EUR
FLUO002840-100G
FLUO002840-500G
Cobalt(III)fluorid
Cobalt(III)fluorid
Formel:
CoF₃ Schmelzpunkt: 927 °C Dichte: 3,88 g/cm³ (20 °C) |
MDL:
MFCD00010942 CAS-Nummer: 10026-18-3 EINECS: 233-062-4 UN: 3260 ADR: 8,II Merck Index: 12,02491 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...