Lithiumhexafluorophosphat

Lieferant: FLUOROCHEM
Gefahr

Synonyme: Lithium hexafluorophosphate(V)

002847-100G
FLUO002847-100GEA 311 EUR
FLUO002847-100G
Lithiumhexafluorophosphat
Lithium hexafluorophosphate
Formel: F₆LiP
Molecular Weight: 151,91 g/mol
Dichte: ∼1,5 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011096
CAS-Nummer: 21324-40-3
EINECS: 244-334-7
UN: 3260
ADR: 8,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR