Tantalpentafluorid
Lieferant: FLUOROCHEM
002867-10G
002867-25G
002867-100G
FLUO002867-10GEA
0
EUR
FLUO002867-10G
FLUO002867-25G
FLUO002867-100G
Tantalpentafluorid
Tantalpentafluorid
Formel:
F₅Ta Molecular Weight: 275.94 g/mol Siedepunkt: 229 Schmelzpunkt: 97 Dichte: 4.74 g/cm³ (20 °C) |
MDL:
MFCD00042542 CAS-Nummer: 7783-71-3 EINECS: 232-022-3 UN: 3260 ADR: 8 II Merck Index: 12,09225 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...