Dibenzo-18-krone-6 (Dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecan, Zinn(II)-Ionophor I)

Lieferant: FLUOROCHEM

Synonyme: Dibenzo-18-crown-6

002907-25G
FLUO002907-25GEA 76.2 EUR
FLUO002907-25G
Dibenzo-18-krone-6 (Dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecan, Zinn(II)-Ionophor I)
Dibenzo-18-krone-6 (Dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecan, Zinn(II)-Ionophor I)
Formel: C₂₀H₂₄O₆
Molecular Weight: 360,41 g/mol
MDL: MFCD00005098
CAS-Nummer: 14187-32-7
EINECS: 238-041-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR