2-(Methylthio)anilin
Lieferant: FLUOROCHEM
002979-25G
FLUO002979-25GEA
55.4
EUR
FLUO002979-25G
2-(Methylthio)anilin
2-(Methylthio)anilin
Formel:
C₇H₉NS Siedepunkt: 62…63 °C (0,05 mmHg) Dichte: 1,1333 g/cm³ (17 °C) |
MDL:
MFCD00007708 CAS-Nummer: 2987-53-3 EINECS: 221-055-9 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...