Thiophen-2-acetonitril

Lieferant: FLUOROCHEM

Synonyme: Thiophene-2-acetonitrile, 2-Thiopheneacetonitrile, 2-(Thiophen-2-yl)acetonitrile

003048-25G
FLUO003048-25GEA 36.9 EUR
FLUO003048-25G
Thiophen-2-acetonitril
Thiophen-2-acetonitril
Formel: C₆H₅NS
Molecular Weight: 123,18 g/mol
MDL: MFCD00005453
CAS-Nummer: 20893-30-5
EINECS: 244-104-6
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR