4'-Hydroxychalkon

Lieferant: FLUOROCHEM

Synonyme: 4'-Hydroxybenzylidenacetophenon

003112-5G 003112-25G
FLUO003112-5GEA 36.9 EUR
FLUO003112-5G FLUO003112-25G
4'-Hydroxychalkon
4'-Hydroxychalkon
Formel: C₁₅H₁₂O₂
Molecular Weight: 224.26 g/mol
MDL: MFCD00016484
CAS-Nummer: 2657-25-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR