4-Fluor-2-(trifluormethyl)anilin
Lieferant: FLUOROCHEM
Synonyme:
2-Amino-α,α,α,5-tetrafluortoluol, α,α,α,4-Tetrafluor-o-toluidin
003127-100G
003127-25G
FLUO003127-100GEA
133
EUR
FLUO003127-100G
FLUO003127-25G
4-Fluor-2-(trifluormethyl)anilin
4-Fluor-2-(trifluormethyl)anilin
Formel:
C₇H₅F₄N Molecular Weight: 179.12 g/mol Siedepunkt: 70 Dichte: 1.38 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00007831 CAS-Nummer: 393-39-5 EINECS: 206-886-7 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...