4-Amino-2,3,5,6-tetrafluorbenzoesäure
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
4-Amino-2,3,5,6-tetrafluorobenzoic Acid
003133-1G
003133-5G
FLUO003133-1GEA
51.8
EUR
FLUO003133-1G
FLUO003133-5G
4-Amino-2,3,5,6-tetrafluorbenzoesäure
4-Amino-2,3,5,6-tetrafluorbenzoesäure
Formel:
C₇H₃F₄NO₂ Molecular Weight: 209.1 g/mol |
MDL:
MFCD00007647 CAS-Nummer: 944-43-4 EINECS: 213-409-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...