1H,1H,7H-Dodecafluor-1-heptanol

Lieferant: FLUOROCHEM

003212-25G 003212-100G 003212-500G
FLUO003212-25GEA 31.1 EUR
FLUO003212-25G FLUO003212-100G FLUO003212-500G
1H,1H,7H-Dodecafluor-1-heptanol
1H,1H,7H-Dodecafluor-1-heptanol
Formel: C₇H₄F₁₂O
MDL: MFCD00039630
CAS-Nummer: 335-99-9
EINECS: 206-406-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR