1H,1H,11H-Eicosafluor-1-unundecanol

Lieferant: FLUOROCHEM

003214-5G 003214-25G
FLUO003214-5GEA 0 EUR
FLUO003214-5G FLUO003214-25G
1H,1H,11H-Eicosafluor-1-unundecanol
1H,1H,11H-Eicosafluor-1-unundecanol
Formel: C₁₁H₄F₂₀O
MDL: MFCD00039628
CAS-Nummer: 307-70-0
EINECS: 206-209-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR