1H,1H,9H-Hexadecafluor-1-nonanol

Lieferant: FLUOROCHEM

003249-25G 003249-100G 003249-250G
FLUO003249-25GEA 31.1 EUR
FLUO003249-25G FLUO003249-100G FLUO003249-250G
1H,1H,9H-Hexadecafluor-1-nonanol
1H,1H,9H-Hexadecafluor-1-nonanol
Formel: C₉H₄F₁₆O
MDL: MFCD00039629
CAS-Nummer: 376-18-1
EINECS: 206-806-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR