Mangan(II)fluorid

Lieferant: FLUOROCHEM

Synonyme: Mangandifluorid

003263-25G 003263.100G
FLUO003263-25GEA 94.8 EUR
FLUO003263-25G FLUO003263.100G
Mangan(II)fluorid
Mangan(II)fluorid
Formel: F₂Mn
Molecular Weight: 92,93 g/mol
Schmelzpunkt: 880…920 °C
Dichte: 3,98 g/cm³ (20 °C)
MDL: MFCD00016222
CAS-Nummer: 7782-64-1
EINECS: 231-960-0
UN: 3288
ADR: 6.1,III
Merck Index: 12,05769

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR