1,3,5-Tris(trifluormethyl)benzol

Lieferant: FLUOROCHEM

003340-1G
FLUO003340-1GEA 84.9 EUR
FLUO003340-1G
1,3,5-Tris(trifluormethyl)benzol
1,3,5-Tris(trifluormethyl)benzol
Formel: C₉H₃F₉
MDL: MFCD00054722
CAS-Nummer: 729-81-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR