6-Methylnicotinsäure
Lieferant: CHEMPUR
Synonyme:
6-Methyl-3-pyridincarbonsäure, 6-Methylpyridin-3-carbonsäure
FL3382.500
003382-1G
003382-5G
003382-25G
CHMPFL3382.500EA
0
EUR
CHMPFL3382.500
FLUO003382-1G
FLUO003382-5G
FLUO003382-25G
6-Methylnicotinsäure
6-Methylnicotinsäure
Formel:
C₇H₇NO₂ Schmelzpunkt: 212 °C |
MDL:
MFCD00006341 CAS-Nummer: 3222-47-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...