2'-Chloracetophenon
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
o-Chloracetophenon, 1-(2-Chlorphenyl)ethanon
003397-25G
FLUO003397-25GEA
36.9
EUR
FLUO003397-25G
2'-Chloracetophenon
2'-Chloracetophenon
Formel:
C₈H₇ClO Molecular Weight: 154,6 g/mol Siedepunkt: 227 °C (1013 hPa) Dichte: 1,189 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000560 CAS-Nummer: 2142-68-9 EINECS: 218-397-6 UN: 3416 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...