3-Indolcarbonsäure

Lieferant: FLUOROCHEM

Synonyme: 1H-Indole-3-carboxylic acid

003408-25G 003408-1G
FLUO003408-25GEA 51.8 EUR
FLUO003408-25G FLUO003408-1G
3-Indolcarbonsäure
3-Indolcarbonsäure
Formel: C₉H₇NO₂
MDL: MFCD00005624
CAS-Nummer: 771-50-6
EINECS: 212-231-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR