3-Amino-1H-1,2,4-triazol-5-thiol

Lieferant: FLUOROCHEM

Synonyme: 3-Amino-5-mercapto-1,2,4-triazole, 3-Amino-1,2,4-triazole-5-thiol, 3-Amino-4H-1,2,4-triazole-5-thiol, 3-Amino-5-mercapto-1,2,4-triazole 5-Amino-4H-1,2,4-triazole-3-thiol

003430-25G
FLUO003430-25GEA 57.9 EUR
FLUO003430-25G
3-Amino-1H-1,2,4-triazol-5-thiol
3-Amino-1H-1,2,4-triazol-5-thiol
Formel: C₂H₄N₄S
Molecular Weight: 116,15 g/mol
MDL: MFCD00005231
CAS-Nummer: 16691-43-3
EINECS: 240-735-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR