(±)-1,2-Dibromhexafluorpropan

Lieferant: FLUOROCHEM

003478-25G 003478-100G
FLUO003478-25GEA 82.1 EUR
FLUO003478-25G FLUO003478-100G
(±)-1,2-Dibromhexafluorpropan
(±)-1,2-Dibromhexafluorpropan
Formel: BrF₂CBrCFCF₃
Molecular Weight: 309.83 g/mol
MDL: MFCD00013568
CAS-Nummer: 661-95-0
EINECS: 211-550-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR