2-Fluorbenzolsulfonylchlorid
Lieferant: FLUOROCHEM
CFF261.10
CFF261.50
003576-5G
003576-25G
CHMPCFF261.10EA
401
EUR
CHMPCFF261.10
CHMPCFF261.50
FLUO003576-5G
FLUO003576-25G
2-Fluorbenzolsulfonylchlorid
2-Fluorbenzolsulfonylchlorid
Formel:
C₆H₄ClFO₂S Siedepunkt: 242 °C (1013 hPa) Dichte: 1,47 g/cm³ (20 °C) |
MDL:
MFCD00042285 CAS-Nummer: 2905-21-7 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...