2,6-Dichlorbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 2,6-Dichlorbenzolsulfonsäurechlorid, 2,6-Dichlorbenzol-1-sulfonylchlorid, 2,6-Dichlorbenzolsulfochlorid

CFF419.10 CFF419.25 CFF419.50 003583-5G
CHMPCFF419.10EA 321 EUR
CHMPCFF419.10 CHMPCFF419.25 CHMPCFF419.50 FLUO003583-5G
2,6-Dichlorbenzolsulfonylchlorid
2,6-Dichlorbenzolsulfonylchlorid
Formel: C₆H₃Cl₃O₂S
Molecular Weight: 245,51 g/mol
Schmelzpunkt: 54…56 °C
MDL: MFCD00052311
CAS-Nummer: 6579-54-0
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR