EEDQ (Ethyl-N-(2-ethoxy-1,2-dihydrochinolin)carboxylat)

Lieferant: FLUOROCHEM

Synonyme: 2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, Ethyl 2-ethoxyquinoline-1(2H)-carboxylate

003625-25G
FLUO003625-25GEA 46.1 EUR
FLUO003625-25G
EEDQ (Ethyl-N-(2-ethoxy-1,2-dihydrochinolin)carboxylat)
EEDQ (Ethyl-N-(2-ethoxy-1,2-dihydrochinolin)carboxylat)
Formel: C₁₄H₁₇NO₃
Molecular Weight: 247.29 g/mol
MDL: MFCD00006703
CAS-Nummer: 16357-59-8
EINECS: 240-418-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR