(4-Hydroxyphenyl)essigsäure
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
4-Hydroxyphenylacetic Acid
003630-25G
003630-100G
FLUO003630-25GEA
42.9
EUR
FLUO003630-25G
FLUO003630-100G
(4-Hydroxyphenyl)essigsäure
(4-Hydroxyphenyl)essigsäure
Formel:
(C₆H₄(OH))CH₂COOH Molecular Weight: 152.15 g/mol Siedepunkt: 347 Schmelzpunkt: 147…150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004347 CAS-Nummer: 156-38-7 EINECS: 205-851-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...