Chlorphenesin
Lieferant: FLUOROCHEM
003642-25G
FLUO003642-25GEA
61.4
EUR
FLUO003642-25G
Chlorphenesin
Chlorphenesin
Formel:
C₉H₁₁ClO₃ Molecular Weight: 202.64 g/mol Schmelzpunkt: 80 |
MDL:
MFCD00021990 CAS-Nummer: 104-29-0 EINECS: 203-192-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...