Allyl-phenylsulfid

Lieferant: FLUOROCHEM

003665-5G
FLUO003665-5GEA 0 EUR
FLUO003665-5G
Allyl-phenylsulfid
Allyl-phenylsulfid
Formel: C₉H₁₀S
Molecular Weight: 150,24 g/mol
MDL: MFCD00014957
CAS-Nummer: 5296-64-0
EINECS: 226-142-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR