(4-Bromphenyl)methylsulfon

Lieferant: FLUOROCHEM

Synonyme: 1-Brom-4-(methylsulfonyl)benzol

003695-5G 003695-25G
FLUO003695-5GEA 35.6 EUR
FLUO003695-5G FLUO003695-25G
(4-Bromphenyl)methylsulfon
(4-Bromphenyl)methylsulfon
Formel: C₇H₇BrO₂S
MDL: MFCD00025065
CAS-Nummer: 3466-32-8
EINECS: 222-421-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR