1,4-Dibrom-2,5-difluorbenzol
Lieferant: FLUOROCHEM
|
Achtung
|
003843-25G
FLUO003843-25GEA
72.9
EUR
FLUO003843-25G
1,4-Dibrom-2,5-difluorbenzol
1,4-Dibromo-2,5-difluorobenzene
Formel:
C₆H₂Br₂F₂ Siedepunkt: 209 °C (1013 hPa) Schmelzpunkt: 65 °C |
MDL:
MFCD00000346 CAS-Nummer: 327-51-5 EINECS: 206-317-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...