2-Chlor-4-(trifluormethyl)anilin
Lieferant: FLUOROCHEM
|
Achtung
|
003965-100G
003965-500G
003965-25G
FLUO003965-100GEA
152
EUR
FLUO003965-100G
FLUO003965-500G
FLUO003965-25G
2-Chlor-4-(trifluormethyl)anilin
2-Chlor-4-(trifluormethyl)anilin
Formel:
C₇H₅ClF₃N Siedepunkt: 89 °C (0,5 mmHg) |
MDL:
MFCD00042563 CAS-Nummer: 39885-50-2 EINECS: 254-674-8 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...