2-Cyan-3-fluoranilin
Lieferant: FLUOROCHEM
Synonyme:
2-Amino-6-fluorbenzonitril, 2-Amino-6-fluorbenzoesäurenitril
003982-25G
003982-100G
FLUO003982-25GEA
34.1
EUR
FLUO003982-25G
FLUO003982-100G
2-Cyan-3-fluoranilin
2-Cyan-3-fluoranilin
Formel:
C₇H₅FN₂ Siedepunkt: 272 °C (1013 hPa) Schmelzpunkt: 127 °C |
MDL:
MFCD00015477 CAS-Nummer: 77326-36-4 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...