4-Amino-2,3,5,6-tetrafluorbenzamid
Lieferant: FLUOROCHEM
004020-1G
FLUO004020-1GEA
31.1
EUR
FLUO004020-1G
4-Amino-2,3,5,6-tetrafluorbenzamid
4-Amino-2,3,5,6-tetrafluorbenzamid
Formel:
C₇H₄F₄N₂O Molecular Weight: 208,12 g/mol |
MDL:
MFCD00008000 CAS-Nummer: 1548-74-9 EINECS: 216-285-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...