1-Brom-3-chlor-5-fluorbenzol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 3-Chloro-5-fluorobromobenzene

004150-25G 004150-100G
FLUO004150-25GEA 34.1 EUR
FLUO004150-25G FLUO004150-100G
1-Brom-3-chlor-5-fluorbenzol
1-Bromo-3-chloro-5-fluorobenzene
Formel: C₆H₃BrClF
Molecular Weight: 209.44 g/mol
MDL: MFCD00070747
CAS-Nummer: 33863-76-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR