1-Brom-4-chlor-3-fluorbenzol
Lieferant: FLUOROCHEM
|
Achtung
|
004152-10G
004152-50G
004152-100G
FLUO004152-10GEA
0
EUR
FLUO004152-10G
FLUO004152-50G
FLUO004152-100G
1-Brom-4-chlor-3-fluorbenzol
1-Brom-4-chlor-3-fluorbenzol
Formel:
C₆H₃BrClF Siedepunkt: 130…142 °C (20 mmHg) Dichte: 1,71 g/cm³ (20 °C) |
MDL:
MFCD00042519 CAS-Nummer: 60811-18-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...