3-Brom-5-chlor-4-iodbenzotrifluorid

Lieferant: FLUOROCHEM

Synonyme: 1-Bromo-3-chloro-2-iodo-5-(trifluoromethyl)benzene

004153-1G
FLUO004153-1GEA 0 EUR
FLUO004153-1G
3-Brom-5-chlor-4-iodbenzotrifluorid
3-Brom-5-chlor-4-iodbenzotrifluorid
Formel: C₇H₂BrClF₃I
MDL: MFCD00042178
CAS-Nummer: 175205-55-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR