2-Brom-3-chlor-5-(trifluormethyl)pyridin
Lieferant: FLUOROCHEM
004164-500MG
FLUO004164-500MGEA
0
EUR
FLUO004164-500MG
2-Brom-3-chlor-5-(trifluormethyl)pyridin
2-Bromo-3-chloro-5-(trifluoromethyl)pyridine
Formel:
C₆H₂BrClF₃N |
MDL:
MFCD00153072 CAS-Nummer: 75806-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...