1-Brom-2,5-dichlor-3-fluorbenzol

Lieferant: FLUOROCHEM
Achtung

004179-5G
FLUO004179-5GEA 39.9 EUR
FLUO004179-5G
1-Brom-2,5-dichlor-3-fluorbenzol
1-Bromo-2,5-dichloro-3-fluorobenzene
Formel: C₆H₂BrCl₂F
Molecular Weight: 243,89 g/mol
MDL: MFCD00142585
CAS-Nummer: 202865-57-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR