4'-Brom-2'-fluoracetanilid
Lieferant: FLUOROCHEM
Synonyme:
N-(4-Brom-2-fluorphenyl)acetamid
004204-25G
004204.100G
FLUO004204-25GEA
91.2
EUR
FLUO004204-25G
FLUO004204.100G
4'-Brom-2'-fluoracetanilid
4'-Brom-2'-fluoracetanilid
Formel:
H₃CC(O)NH(C₆H₃BrF) Molecular Weight: 232,05 g/mol Siedepunkt: 344 °C (1013 hPa) Schmelzpunkt: 158…160 °C |
MDL:
MFCD00042588 CAS-Nummer: 326-66-9 EINECS: 206-314-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...