3-Brom-4-fluortoluol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
1-Bromo-2-fluoro-5-methylbenzene
004234-10G
004234-100G
004234-25G
FLUO004234-10GEA
0
EUR
FLUO004234-10G
FLUO004234-100G
FLUO004234-25G
3-Brom-4-fluortoluol
3-Brom-4-fluortoluol
Formel:
C₇H₆BrF Molecular Weight: 189.03 g/mol Siedepunkt: 186 Dichte: 1.5 g/cm³ (20 °C) |
MDL:
MFCD00040827 CAS-Nummer: 452-62-0 EINECS: 207-201-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...