4'-Brom-2'-(trifluormethoxy)acetanilid

Lieferant: FLUOROCHEM

004297-5G
FLUO004297-5GEA 48.8 EUR
FLUO004297-5G
4'-Brom-2'-(trifluormethoxy)acetanilid
4'-Brom-2'-(trifluormethoxy)acetanilid
Formel: C₉H₇BrF₃NO₂
MDL: MFCD00203979
CAS-Nummer: 175278-18-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR