2-Chlor-4-(trifluormethyl)benzonitril

Lieferant: FLUOROCHEM

004384-1G 004384-25G 004384-5G
FLUO004384-1GEA 0 EUR
FLUO004384-1G FLUO004384-25G FLUO004384-5G
2-Chlor-4-(trifluormethyl)benzonitril
2-Chlor-4-(trifluormethyl)benzonitril
Formel: C₈H₃ClF₃N
MDL: MFCD00084939
CAS-Nummer: 1813-33-8
UN: 3276
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR