1-Chlor-2,6-dibrom-4-fluorbenzol
Lieferant: FLUOROCHEM
Synonyme:
2-Chlor-1,3-dibrom-5-fluorbenzol
004387-25G
FLUO004387-25GEA
69.8
EUR
FLUO004387-25G
1-Chlor-2,6-dibrom-4-fluorbenzol
1-Chloro-2,6-dibromo-4-fluorobenzene
Formel:
C₆H₂Br₂ClF Molecular Weight: 288,34 g/mol |
MDL:
MFCD00061122 CAS-Nummer: 179897-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...