2-Chlor-2,2-difluoracetylchlorid
Lieferant: FLUOROCHEM
004393-5G
FLUO004393-5GEA
0
EUR
FLUO004393-5G
2-Chlor-2,2-difluoracetylchlorid
2-Chlor-2,2-difluoracetylchlorid
Formel:
C₂Cl₂F₂O |
MDL:
MFCD00039317 CAS-Nummer: 354-24-5 EINECS: 206-551-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...