3'-Chlor-4'-fluoracetophenon

Lieferant: FLUOROCHEM

004415-5G
FLUO004415-5GEA 0 EUR
FLUO004415-5G
3'-Chlor-4'-fluoracetophenon
3'-Chlor-4'-fluoracetophenon
Formel: C₈H₆ClFO
MDL: MFCD00042203
CAS-Nummer: 2923-66-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR