4-Chlor-3-fluoranisol
Lieferant: FLUOROCHEM
Synonyme:
1-Chloro-2-fluoro-4-methoxybenzene
004420-5G
004420-10G
004420-50G
004420-25G
FLUO004420-5GEA
69.8
EUR
FLUO004420-5G
FLUO004420-10G
FLUO004420-50G
FLUO004420-25G
4-Chlor-3-fluoranisol
4-Chlor-3-fluoranisol
Formel:
C₇H₆ClFO Molecular Weight: 160.58 g/mol Siedepunkt: 196 Dichte: 1.26 g/cm³ (20 °C) |
MDL:
MFCD00042570 CAS-Nummer: 501-29-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...