3-Chlor-4-fluorbenzamid
Lieferant: FLUOROCHEM
Synonyme:
3-Chlor-4-fluorbenzoesäureamid
CFF923.
004424-5G
004424-25G
CHMPCFF923.EA
0
EUR
CHMPCFF923.
FLUO004424-5G
FLUO004424-25G
3-Chlor-4-fluorbenzamid
3-Chlor-4-fluorbenzamid
Formel:
C₇H₅ClFNO Molecular Weight: 173.57 g/mol |
MDL:
MFCD00153092 CAS-Nummer: 701-43-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...