3-Chlor-4-fluorbenzamid

Lieferant: FLUOROCHEM

Synonyme: 3-Chlor-4-fluorbenzoesäureamid

CFF923. 004424-5G 004424-25G
CHMPCFF923.EA 0 EUR
CHMPCFF923. FLUO004424-5G FLUO004424-25G
3-Chlor-4-fluorbenzamid
3-Chlor-4-fluorbenzamid
Formel: C₇H₅ClFNO
Molecular Weight: 173.57 g/mol
MDL: MFCD00153092
CAS-Nummer: 701-43-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR