3-Chlor-4-fluorbenzonitril

Lieferant: FLUOROCHEM

Synonyme: 1-Chlor-3-cyan-6-fluorbenzene

CFF940. 004434-25G 004434-100G
CHMPCFF940.EA 0 EUR
CHMPCFF940. FLUO004434-25G FLUO004434-100G
3-Chlor-4-fluorbenzonitril
3-Chlor-4-fluorbenzonitril
Formel: C₇H₃ClFN
Molecular Weight: 155.56 g/mol
Siedepunkt: 135 °C (65 mmHg)
Schmelzpunkt: 70…71 °C
MDL: MFCD00015431
CAS-Nummer: 117482-84-5
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR