2-Chlor-4-fluorbenzylamin

Lieferant: FLUOROCHEM

004441-1G 004441-5G 004441-25G
FLUO004441-1GEA 0 EUR
FLUO004441-1G FLUO004441-5G FLUO004441-25G
2-Chlor-4-fluorbenzylamin
2-Chlor-4-fluorbenzylamin
Formel: C₇H₇ClFN
Molecular Weight: 159.59 g/mol
Siedepunkt: 94…95 °C (15 mmHg)
MDL: MFCD00042532
CAS-Nummer: 15205-11-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR